Assay Details
| Gene Symbol: ISG20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258053.1 | NP_001244982.1 | 734 | 76 | ||
| XM_028850241.1 | XP_028706074.1 | 735 | 76 | |||
| GenBank mRNA | JU473329.1 | - | 734 | 76 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ISG20
Genomic Map