Assay Details
| Gene Symbol: SMG9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194356.2 | NP_001181285.1 | 718 | 119 | ||
| XM_015124116.2 | XP_014979602.1 | 882 | 119 | |||
| XM_015124117.2 | XP_014979603.1 | 873 | 119 | |||
| XM_015124118.2 | XP_014979604.1 | 882 | 119 | |||
| XM_015124119.2 | XP_014979605.1 | 882 | 119 | |||
| XM_015124120.2 | XP_014979606.1 | 882 | 119 | |||
| GenBank mRNA | JU327272.1 | - | 438 | 119 | ||
| JU476414.1 | - | 765 | 119 | |||
| JV047657.1 | - | 770 | 119 | |||
| JV689934.1 | - | 608 | 119 | |||
More Information
Set Membership: |
|

SMG9
Genomic Map