Assay Details
| Gene Symbol: KCNG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148781.2 | XP_015004267.1 | 1078 | 80 | ||
| GenBank mRNA | JV650789.1 | - | 906 | 80 | ||
| JV683269.1 | - | 1053 | 80 | |||
More Information
Set Membership: |
|

KCNG1
Genomic Map