Assay Details
| Gene Symbol: ACVR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260761.1 | NP_001247690.1 | 1259 | 98 | ||
| XM_015110144.2 | XP_014965630.1 | 1795 | 98 | |||
| XM_015110145.2 | XP_014965631.1 | 1370 | 98 | |||
| XM_015110146.2 | XP_014965632.1 | 1910 | 98 | |||
| GenBank mRNA | JU319950.1 | - | 1312 | 98 | ||
| JU472698.1 | - | 1245 | 98 | |||
| JV043497.1 | - | 1259 | 98 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ACVR1
Genomic Map