Assay Details
| Gene Symbol: KLHL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195300.2 | NP_001182229.1 | 678 | 83 | ||
| XM_028837138.1 | XP_028692971.1 | 764 | 83 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KLHL1| Gene Symbol: KLHL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195300.2 | NP_001182229.1 | 678 | 83 | ||
| XM_028837138.1 | XP_028692971.1 | 764 | 83 | |||
Set Membership: |
|