Assay Details
| Gene Symbol: EMC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001085740.4 | XP_001085740.4 | 679 | 99 | ||
| GenBank mRNA | JU335512.1 | - | 590 | 99 | ||
| JU476452.1 | - | 589 | 99 | |||
| JV066077.1 | - | 572 | 99 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

EMC7
Genomic Map