Assay Details
| Gene Symbol: PRKAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266032.1 | NP_001252961.1 | 942 | 73 | ||
| XM_028828927.1 | XP_028684760.1 | 2965 | 73 | |||
| XM_028828928.1 | XP_028684761.1 | 888 | 73 | |||
| XM_028828929.1 | XP_028684762.1 | 923 | 73 | |||
| GenBank mRNA | JU320851.1 | - | 943 | 73 | ||
| JU471484.1 | - | 911 | 73 | |||
| JV045737.1 | - | 942 | 73 | |||
| JV452056.1 | - | 938 | 73 | |||
More Information
Set Membership: |
|

PRKAG1
Genomic Map