Assay Details
| Gene Symbol: HTATIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001093563.4 | XP_001093563.2 | 631 | 142 | ||
| XM_015114635.2 | XP_014970121.1 | 525 | 142 | |||
| XM_015114636.2 | XP_014970122.1 | 547 | 142 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HTATIP2
Genomic Map