Assay Details
| Gene Symbol: STX18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260531.1 | NP_001247460.1 | 851 | 89 | ||
| XM_028848103.1 | XP_028703936.1 | 884 | 89 | |||
| XM_028848104.1 | XP_028703937.1 | 952 | 89 | |||
| GenBank mRNA | JU341815.1 | - | 3398 | 89 | ||
| JU476028.1 | - | 893 | 89 | |||
| JV047396.1 | - | 851 | 89 | |||
More Information
Set Membership: |
|

STX18
Genomic Map