Assay Details
| Gene Symbol: TIMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032939.1 | NP_001028111.1 | 518 | 75 | ||
| GenBank mRNA | AF366397.1 | - | 518 | 75 | ||
| JU336136.1 | - | 635 | 75 | |||
| JU476998.1 | - | 637 | 75 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TIMP1
Genomic Map