Assay Details
| Gene Symbol: FZD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265729.1 | NP_001252658.1 | 1595 | 62 | ||
| XM_015145733.2 | XP_015001219.1 | 1643 | 62 | |||
| XM_015145734.2 | XP_015001220.1 | 1486 | 62 | |||
| XM_028852118.1 | XP_028707951.1 | 1420 | 62 | |||
| GenBank mRNA | JU326838.1 | - | 951 | 62 | ||
| JV636398.1 | - | 1595 | 62 | |||
More Information
Set Membership: |
|

FZD6
Genomic Map