Assay Details
| Gene Symbol: CS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151991.2 | XP_015007477.1 | 474 | 88 | ||
| XM_015151992.2 | XP_015007478.1 | 308 | 88 | |||
| XM_028829611.1 | XP_028685444.1 | 303 | 88 | |||
| GenBank mRNA | JU333037.1 | - | 447 | 88 | ||
| JU474381.1 | - | 446 | 88 | |||
| JV046243.1 | - | 464 | 88 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CS
Genomic Map