Assay Details
| Gene Symbol: NT5C1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001113402.4 | XP_001113402.3 | 764 | 55 | ||
| XM_015135914.2 | XP_014991400.2 | 771 | 55 | |||
| GenBank mRNA | JV560493.1 | - | 77 | 55 | ||
More Information
Set Membership: |
|

NT5C1A
Genomic Map