Assay Details
| Gene Symbol: PAK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123893.2 | XP_014979379.2 | 1532 | 58 | ||
| XM_015123894.2 | XP_014979380.2 | 1532 | 58 | |||
| XM_015123895.2 | XP_014979381.2 | 1070 | 58 | |||
| XM_028839953.1 | XP_028695786.1 | 1550 | 58 | |||
| GenBank mRNA | JV635906.1 | - | 1404 | 58 | ||
| JV640026.1 | - | 1477 | 58 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PAK4
Genomic Map