Assay Details
| Gene Symbol: FUS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001112691.4 | XP_001112691.2 | 2377 | 69 | ||
| XM_015125995.2 | XP_014981481.2 | 2392 | 69 | |||
| XM_015125996.2 | XP_014981482.2 | 2374 | 69 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FUS
Genomic Map