Assay Details
| Gene Symbol: PIGH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107223.4 | XP_001107223.2 | 287 | 56 | ||
| XM_028852044.1 | XP_028707877.1 | 288 | 56 | |||
| XM_028852045.1 | XP_028707878.1 | 288 | 56 | |||
| XM_028852046.1 | XP_028707879.1 | 290 | 56 | |||
| XM_028852047.1 | XP_028707880.1 | 288 | 56 | |||
| GenBank mRNA | JU346180.1 | - | 2711 | 56 | ||
More Information
Set Membership: |
|

PIGH
Genomic Map