Assay Details
| Gene Symbol: STX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257724.1 | NP_001244653.1 | 421 | 104 | ||
| GenBank mRNA | JU322469.1 | - | 421 | 104 | ||
| JV566428.1 | - | 436 | 104 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

STX7
Genomic Map