Assay Details
Gene Symbol: ARFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002799671.2 | XP_002799717.1 | 790 | 70 | ||
XM_015114973.2 | XP_014970459.1 | 1061 | 70 | |||
XM_015114974.2 | XP_014970460.1 | 941 | 70 | |||
XM_015114975.1 | XP_014970461.1 | 439 | 70 | |||
GenBank mRNA | JU320506.1 | - | 707 | 70 | ||
JU473592.1 | - | 701 | 70 | |||
JV044477.1 | - | 755 | 70 | |||
JV637699.1 | - | 712 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
