Assay Details
| Gene Symbol: SLC1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261669.1 | NP_001248598.1 | 1446 | 100 | ||
| XM_015114491.2 | XP_014969977.1 | 1067 | 100 | |||
| XM_015114492.2 | XP_014969978.1 | 950 | 100 | |||
| XM_015114493.2 | XP_014969979.1 | 983 | 100 | |||
| XM_028832500.1 | XP_028688333.1 | 908 | 100 | |||
| GenBank mRNA | JU319738.1 | - | 1446 | 100 | ||
| JV529720.1 | - | 1452 | 100 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC1A2
Genomic Map