Assay Details
Gene Symbol: WDR53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015132591.2 | XP_014988077.1 | 645 | 73 | ||
XM_028843537.1 | XP_028699370.1 | 708 | 73 | |||
XM_028843538.1 | XP_028699371.1 | 592 | 73 | |||
GenBank mRNA | JU324114.1 | - | 497 | 73 | ||
JU485838.1 | - | 375 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
