Assay Details
| Gene Symbol: MYOZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139140.2 | XP_014994626.2 | 458 | 122 | ||
| GenBank mRNA | JV063245.1 | - | 389 | 122 | ||
| JV653621.1 | - | 351 | 122 | |||
More Information
Set Membership: |
|

MYOZ2
Genomic Map