Assay Details
| Gene Symbol: PIGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260603.1 | NP_001247532.1 | 833 | 150 | ||
| XM_015126955.2 | XP_014982441.1 | 948 | 150 | |||
| XM_015126956.2 | XP_014982442.1 | 1264 | 150 | |||
| GenBank mRNA | JU334996.1 | - | 795 | 150 | ||
| JU471631.1 | - | 833 | 150 | |||
More Information
Set Membership: |
|

PIGA
Genomic Map