Assay Details
| Gene Symbol: RTN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205127.2 | NP_001192056.2 | 3365 | 90 | ||
| XM_015112348.2 | XP_014967834.1 | 2924 | 90 | |||
| XM_015112349.2 | XP_014967835.2 | 2226 | 90 | |||
| XM_015112350.2 | XP_014967836.1 | 762 | 90 | |||
| GenBank mRNA | CK231727.1 | - | 193 | 90 | ||
| JU471546.1 | - | 580 | 90 | |||
| JV045048.1 | - | 474 | 90 | |||
| JV588048.1 | - | 3582 | 90 | |||
| JV622917.1 | - | 3262 | 90 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RTN4
Genomic Map