Assay Details
| Gene Symbol: SLC6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257768.1 | NP_001244697.1 | 1626 | 68 | ||
| XM_015130223.2 | XP_014985709.1 | 1905 | 68 | |||
| GenBank mRNA | JU319578.1 | - | 1626 | 68 | ||
More Information
Set Membership: |
|

SLC6A1
Genomic Map