Assay Details
| Gene Symbol: LHX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261792.1 | NP_001248721.1 | 150 | 64 | ||
| GenBank mRNA | JU337139.1 | - | 150 | 64 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LHX2| Gene Symbol: LHX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261792.1 | NP_001248721.1 | 150 | 64 | ||
| GenBank mRNA | JU337139.1 | - | 150 | 64 | ||
Set Membership: |
|