Assay Details
| Gene Symbol: LHPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001083060.4 | XP_001083060.2 | 797 | 81 | ||
| GenBank mRNA | JU322814.1 | - | 774 | 81 | ||
| JU471641.1 | - | 797 | 81 | |||
| JV044427.1 | - | 752 | 81 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LHPP
Genomic Map