Assay Details
| Gene Symbol: LOC114675369 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028842628.1 | XP_028698461.1 | 257 | 85 | ||
| Gene Symbol: TLL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139468.2 | XP_014994954.1 | 2186 | 85 | ||
| XM_028848870.1 | XP_028704703.1 | 2292 | 85 | |||
More Information
Set Membership: |
|

LOC114675369
Genomic Map