Assay Details
| Gene Symbol: VAMP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257789.1 | NP_001244718.1 | 180 | 114 | ||
| GenBank mRNA | JU320802.1 | - | 180 | 114 | ||
| JU473789.1 | - | 246 | 114 | |||
| JV045572.1 | - | 256 | 114 | |||
| JV639417.1 | - | 288 | 114 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

VAMP8
Genomic Map