Assay Details
| Gene Symbol: RPIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258126.1 | NP_001245055.1 | 624 | 64 | ||
| GenBank mRNA | JU330972.1 | - | 500 | 64 | ||
| JU474756.1 | - | 624 | 64 | |||
| JV045203.1 | - | 638 | 64 | |||
More Information
Set Membership: |
|

RPIA
Genomic Map