Assay Details
| Gene Symbol: LGALSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112578.2 | XP_014968064.2 | 647 | 56 | ||
| GenBank mRNA | JU475286.1 | - | 276 | 56 | ||
| JV046844.1 | - | 205 | 56 | |||
More Information
Set Membership: |
|

LGALSL
Genomic Map