Assay Details
| Gene Symbol: SLC20A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266611.1 | NP_001253540.1 | 710 | 67 | ||
| XM_015145238.2 | XP_015000724.1 | 3179 | 67 | |||
| XM_015145239.2 | XP_015000725.1 | 872 | 67 | |||
| XM_015145241.2 | XP_015000727.1 | 1100 | 67 | |||
| XM_015145242.2 | XP_015000728.1 | 3336 | 67 | |||
| XM_015145243.2 | XP_015000729.1 | 816 | 67 | |||
| XM_015145244.2 | XP_015000730.1 | 819 | 67 | |||
| XM_015145246.2 | XP_015000732.1 | 3304 | 67 | |||
| XM_028852239.1 | XP_028708072.1 | 931 | 67 | |||
| XM_028852240.1 | XP_028708073.1 | 822 | 67 | |||
| GenBank mRNA | JU336009.1 | - | 703 | 67 | ||
| JV047986.1 | - | 710 | 67 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015145246.2 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC20A2
Genomic Map