Assay Details
| Gene Symbol: CADM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124158.2 | XP_014979644.1 | 1144 | 67 | ||
| XM_028839256.1 | XP_028695089.1 | 993 | 67 | |||
| GenBank mRNA | JU327819.1 | - | 884 | 67 | ||
| JV602304.1 | - | 1096 | 67 | |||
| JV667242.1 | - | 1083 | 67 | |||
More Information
Set Membership: |
|

CADM4
Genomic Map