Assay Details
| Gene Symbol: SCN2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100011.4 | XP_001100011.1 | 2281 | 107 | ||
| XM_001100368.4 | XP_001100368.1 | 750 | 107 | |||
| XM_015110269.2 | XP_014965755.1 | 757 | 107 | |||
| XM_028830788.1 | XP_028686621.1 | 523 | 107 | |||
| GenBank mRNA | JU324497.1 | - | 748 | 107 | ||
| JU324527.1 | - | 748 | 107 | |||
| JV516417.1 | - | 752 | 107 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SCN2A
Genomic Map