Assay Details
| Gene Symbol: FIZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125140.2 | XP_014980626.1 | 1413 | 143 | ||
| XM_015125141.2 | XP_014980627.1 | 451 | 143 | |||
| XM_028839854.1 | XP_028695687.1 | 663 | 143 | |||
| GenBank mRNA | JU322971.1 | - | 390 | 143 | ||
More Information
Set Membership: |
|

FIZ1
Genomic Map