Assay Details
| Gene Symbol: STK25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015111555.2 | XP_014967041.1 | 616 | 62 | ||
| XM_015111557.2 | XP_014967043.1 | 552 | 62 | |||
| XM_028831361.1 | XP_028687194.1 | 512 | 62 | |||
| XM_028831362.1 | XP_028687195.1 | 477 | 62 | |||
| XM_028831363.1 | XP_028687196.1 | 577 | 62 | |||
| GenBank mRNA | JU319641.1 | - | 365 | 62 | ||
| JV045926.1 | - | 370 | 62 | |||
| JV492112.1 | - | 370 | 62 | |||
| JV603553.1 | - | 457 | 62 | |||
More Information
Set Membership: |
|

STK25
Genomic Map