Assay Details
| Gene Symbol: SUPT16H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143253.2 | XP_014998739.1 | 2384 | 67 | ||
| GenBank mRNA | JU332735.1 | - | 2399 | 67 | ||
| JU474157.1 | - | 2437 | 67 | |||
| JV044607.1 | - | 2473 | 67 | |||
More Information
Set Membership: |
|

SUPT16H
Genomic Map