Assay Details
| Gene Symbol: NEDD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152491.2 | XP_015007977.1 | 1437 | 76 | ||
| XM_015152493.2 | XP_015007979.1 | 1900 | 76 | |||
| XM_015152494.2 | XP_015007980.1 | 1402 | 76 | |||
| XM_015152495.2 | XP_015007981.1 | 1402 | 76 | |||
| XM_028829793.1 | XP_028685626.1 | 1080 | 76 | |||
More Information
Set Membership: |
|

NEDD1
Genomic Map