Assay Details
| Gene Symbol: ELF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260954.2 | NP_001247883.1 | 1039 | 111 | ||
| XM_015120889.2 | XP_014976375.1 | 1360 | 111 | |||
| XM_015120891.2 | XP_014976377.1 | 1258 | 111 | |||
| GenBank mRNA | JU333294.1 | - | 1044 | 111 | ||
| JU474608.1 | - | 1155 | 111 | |||
| JV046388.1 | - | 1042 | 111 | |||
More Information
Set Membership: |
|

ELF1
Genomic Map