Assay Details
| Gene Symbol: MARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266948.1 | NP_001253877.1 | 1663 | 67 | ||
| XM_015152050.2 | XP_015007536.2 | 1780 | 67 | |||
| XM_015152051.2 | XP_015007537.2 | 1535 | 67 | |||
| XM_028828997.1 | XP_028684830.1 | 1780 | 67 | |||
| GenBank mRNA | JU322390.1 | - | 1648 | 67 | ||
| JU471246.1 | - | 1663 | 67 | |||
| JV044407.1 | - | 1637 | 67 | |||
More Information
Set Membership: |
|

MARS1
Genomic Map