Assay Details
| Gene Symbol: SP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266828.1 | NP_001253757.1 | 2126 | 68 | ||
| GenBank mRNA | JU336058.1 | - | 2258 | 68 | ||
| JU476917.1 | - | 2126 | 68 | |||
More Information
Set Membership: |
|

SP4
Genomic Map