Assay Details
| Gene Symbol: YWHAG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194436.3 | NP_001181365.1 | 332 | 71 | ||
| GenBank mRNA | JU477101.1 | - | 292 | 71 | ||
| JV043828.1 | - | 316 | 71 | |||
| JV635820.1 | - | 309 | 71 | |||
More Information
Set Membership: |
|

YWHAG
Genomic Map