Assay Details
| Gene Symbol: MAPKAPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001085549.4 | XP_001085549.2 | 757 | 63 | ||
| XM_015118000.2 | XP_014973486.1 | 757 | 63 | |||
| GenBank mRNA | JU328604.1 | - | 333 | 63 | ||
| JV665650.1 | - | 352 | 63 | |||
More Information
Set Membership: |
|

MAPKAPK2
Genomic Map