Assay Details
| Gene Symbol: CTSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001108862.4 | XP_001108862.2 | 412 | 153 | ||
| GenBank mRNA | JU481072.1 | - | 197 | 153 | ||
| JV643372.1 | - | 286 | 153 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CTSH
Genomic Map