Assay Details
| Gene Symbol: ORMDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266349.1 | NP_001253278.1 | 279 | 81 | ||
| XM_015110498.2 | XP_014965984.1 | 1499 | 81 | |||
| XM_015110499.2 | XP_014965985.1 | 401 | 81 | |||
| XM_015110500.2 | XP_014965986.1 | 309 | 81 | |||
| XM_028830555.1 | XP_028686388.1 | 459 | 81 | |||
| XM_028830556.1 | XP_028686389.1 | 428 | 81 | |||
| GenBank mRNA | JU320839.1 | - | 278 | 81 | ||
| JU477898.1 | - | 279 | 81 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ORMDL1
Genomic Map