Assay Details
| Gene Symbol: TNFRSF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113309.2 | XP_014968795.2 | 1273 | 94 | ||
| XM_015113371.2 | XP_014968857.2 | 1306 | 94 | |||
More Information
Set Membership: |
|

TNFRSF8
Genomic Map