Assay Details
| Gene Symbol: EPRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116029.2 | XP_014971515.1 | 4886 | 71 | ||
| XM_015116038.2 | XP_014971524.1 | 4865 | 71 | |||
| GenBank mRNA | JU474647.1 | - | 4496 | 71 | ||
| JV046429.1 | - | 4511 | 71 | |||
| JV565078.1 | - | 4426 | 71 | |||
| JV709245.1 | - | 4510 | 71 | |||
More Information
Set Membership: |
|

EPRS1
Genomic Map