Assay Details
| Gene Symbol: PTGDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193930.2 | NP_001180859.1 | 203 | 57 | ||
| GenBank mRNA | JU476517.1 | - | 192 | 57 | ||
| JV047747.1 | - | 202 | 57 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PTGDS
Genomic Map