Assay Details
| Gene Symbol: CA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266383.1 | NP_001253312.1 | 830 | 67 | ||
| GenBank mRNA | JU320654.1 | - | 830 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CA10| Gene Symbol: CA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266383.1 | NP_001253312.1 | 830 | 67 | ||
| GenBank mRNA | JU320654.1 | - | 830 | 67 | ||
Set Membership: |
|