Assay Details
| Gene Symbol: AZI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266975.1 | NP_001253904.1 | 782 | 142 | ||
| XM_015132273.2 | XP_014987759.2 | 854 | 142 | |||
| GenBank mRNA | JU335914.1 | - | 829 | 142 | ||
| JU476791.1 | - | 714 | 142 | |||
| JV047926.1 | - | 782 | 142 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AZI2
Genomic Map