Assay Details
| Gene Symbol: LOC106993830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028842748.1 | XP_028698581.1 | 928 | 130 | ||
| XM_028842749.1 | XP_028698582.1 | 903 | 130 | |||
| XM_028842750.1 | XP_028698583.1 | 903 | 130 | |||
| XM_028842751.1 | XP_028698584.1 | 903 | 130 | |||
| XM_028842752.1 | XP_028698585.1 | 903 | 130 | |||
| Gene Symbol: SECISBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090428.4 | XP_001090428.2 | 2103 | 130 | ||
| XM_001090657.3 | XP_001090657.2 | 2219 | 130 | |||
| XM_015118054.2 | XP_014973540.2 | 2104 | 130 | |||
| XM_015118055.2 | XP_014973541.2 | 2101 | 130 | |||
| XM_015118058.2 | XP_014973544.2 | 2636 | 130 | |||
| XM_028835315.1 | XP_028691148.1 | 2219 | 130 | |||
| XM_028835316.1 | XP_028691149.1 | 2757 | 130 | |||
| XM_028835317.1 | XP_028691150.1 | 2496 | 130 | |||
| GenBank mRNA | JU336342.1 | - | 2192 | 130 | ||
| JV723171.1 | - | 2194 | 130 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LOC106993830
Genomic Map